Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQSEC2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB164085 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164085 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164085 Supplier Novus Biologicals Supplier No. NBP281723JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IQSEC2 Polyclonal antibody specifically detects IQSEC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen IQSEC2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias BRAG1, brefeldin A resistant Arf-guanine nucleotide exchange factor 1, brefeldin A resistant Arf-guanine nucleotide exchange factor 1c, IQ motif and Sec7 domain 2, IQ motif and SEC7 domain-containing protein 2, KIAA0522, MRX1
Host Species Rabbit
Immunogen IQSEC2 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human IQSEC2. The immunogen is located within amino acids 1380 - 1430 of IQSEC2. Amino Acid Squence: PGSRPPGGSYSHPHHPQS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23096
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.