Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177068SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177068SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177068SS Supplier Novus Biologicals Supplier No. NBP177068SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRAK1 Polyclonal specifically detects IRAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen IRAK1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-4 ug/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 ug/mL, Immunoprecipitation 1:10-1:500
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P51617
Gene Alias EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle
Gene Symbols IRAK1
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the carboxy terminus of human IRAK. The immunogen is located within the last 50 amino acids of IRAK. Amino Acid Squence: DRQGPEESDEFQS
Molecular Weight of Antigen 77 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3654
Test Specificity At least four isoforms of IRAK1 are known to exist; this antibody will detect all four isoforms. IRAK1 antibody is predicted to not cross-react with other members of the IRAK protein family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.