Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB357704 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357704 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357704 Supplier Novus Biologicals Supplier No. NBP176641MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRAK2 Polyclonal antibody specifically detects IRAK2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen IRAK2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias interleukin-1 receptor associated kinase-2, interleukin-1 receptor-associated kinase 2, interleukin-1 receptor-associated kinase-like 2, IRAK-2, MGC150550
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of mouse IRAK2a. The immunogen is located within the last 50 amino acids of IRAK2. Amino Acid Squence: EAKRRLMENIVLYKEER
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3656
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.