Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176782 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176782 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176782 Supplier Novus Biologicals Supplier No. NBP176782
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRAK3 Polyclonal specifically detects IRAK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen IRAK3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 2 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. AF113136
Gene Alias ASRT5, EC 2.7.11.1, FLJ13601, IL-1 receptor-associated kinase M, interleukin-1 receptor-associated kinase 3, IRAK-3, IRAKM, IRAK-M
Gene Symbols IRAK3
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 16 amino acids near the carboxy terminus of human IRAK-M. The immunogen is located within the last 50 amino acids of IRAK-M. Amino Acid Squence: CSSKFSWDEYEQYKKE
Molecular Weight of Antigen 70 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 11213
Test Specificity IRAK-M antibody is predicted to have no cross reactivity to other members of the IRAK family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.