Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK4 Antibody (413613), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB371259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371259 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371259 Supplier Novus Biologicals Supplier No. FAB39191D
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IRAK4 Monoclonal specifically detects IRAK4 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry.

Specifications

Antigen IRAK4
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 413613
Conjugate DyLight 350
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64
Host Species Rat
Immunogen E. coli-derived recombinant human IRAK4, Met1-Ser460, Accession # Q9NWZ3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 51135
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.