Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK4 Antibody (6F8), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. DDX0340A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0340A647 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0340A647 Supplier Novus Biologicals Supplier No. DDX0340A647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IRAK4 Monoclonal specifically detects IRAK4 in Canine, Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IRAK4
Applications Flow Cytometry
Classification Monoclonal
Clone 6F8
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 1:10-1:1000
Gene Alias EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64
Gene Symbols IRAK4
Host Species Mouse
Immunogen IRAK-4 was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 51135
Target Species Canine, Human
Content And Storage Store at -20C in the dark. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.