Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK4 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB218356 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB218356 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB218356 Supplier Novus Biologicals Supplier No. NBP177231AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRAK4 Polyclonal antibody specifically detects IRAK4 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen IRAK4
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 13 amino acids near the carboxy terminus of human IRAK-4. The immunogen is located within the last 50 amino acids of IRAK-4. Amino Acid Squence: KKVQQLLQEMTAS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 51135
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.