Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK4 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB324907 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324907 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324907 Supplier Novus Biologicals Supplier No. NBP299534MFV500

Rabbit Polyclonal Antibody

IRAK4 Polyclonal antibody specifically detects IRAK4 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen IRAK4
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human IRAK4 (Uniprot#: Q9NWZ3-1; Met1-Ser460)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 51135
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.