Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK4 Antibody, Novus Biologicals™
SDP

Catalog No. NB50059701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50059701 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50059701 Supplier Novus Biologicals Supplier No. NB5005970.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 12 publications

IRAK4 Polyclonal specifically detects IRAK4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.

Specifications

Antigen IRAK4
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000, Flow Cytometry, Immunoprecipitation
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. NP_001107654
Gene Alias EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64
Gene Symbols IRAK4
Host Species Rabbit
Immunogen This antibody was developed against a mixture of synthetic peptides containing amino acids 38-54 and 120-136 of mouse IRAK-4.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 51135
Test Specificity Cross-reacts with Human and Mouse.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.