Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRE1 alpha Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB1002324B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002324B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002324B Supplier Novus Biologicals Supplier No. NB1002324B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRE1 alpha Polyclonal specifically detects IRE1 alpha in Human, Mouse, Rat samples. It is validated for Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Western Blot.

Specifications

Antigen IRE1 alpha
Applications Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O75460
Gene Alias EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, IRE1a, Ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1
Gene Symbols ERN1
Host Species Rabbit
Immunogen This IRE1 alpha antibody was raised against a synthetic peptide within the human IRE1 alpha protein (within residues 700-800). [Swiss-Prot #O75460]
Molecular Weight of Antigen 110 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Neuroscience, Signal Transduction, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 2081
Test Specificity This antibody detects the levels of endogenous total IRE1 Alpha protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.