Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRE1 alpha, p Ser724 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002323 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002323 0.1 mL
NB1002323SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002323 Supplier Novus Biologicals Supplier No. NB1002323
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 252 publications

IRE1 alpha (p Ser724) Polyclonal specifically detects IRE1 alpha (p Ser724) in Drosophila, Goat, Golden Syrian Hamster, Human, Mammalia, Monkey, Mouse, Pig, Primate, Rabbit, Rat samples. It is validated for ChIP Assay, ELISA, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown, Simple Western, Western Blot.

Specifications

Antigen IRE1 alpha (p Ser724)
Applications ChIP Assay, ELISA, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoblotting, In vitro assay, Chromatin Immunoprecipitation (ChIP), KnockDown Validated
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O75460
Gene Alias EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, ire1 alpha p Ser724, ire1 alpha phosphorylation, IRE1a, ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, phospho ire1 alpha, phospho-ire1 alpha, phospho-ire1a, p-ire1 alpha, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1
Gene Symbols ERN1
Host Species Rabbit
Immunogen This IRE1 alpha [p Ser724] antibody was raised against a synthetic peptide surrounding the phosphorylated serine 724 of the human IRE1 alpha protein. [Swiss-Prot #O75460]
Molecular Weight of Antigen 110 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Neuroscience, Phospho Specific, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 2081
Test Specificity NB100-2323 IRE1 alpha [p Ser724] Antibody detects IRE-1 alpha when phosphorylated at Ser724 residue.
Target Species Drosophila, Mouse, Goat, Golden Syrian Hamster, Human, Goat, Mammalia, Monkey, Monkey, Rabbit, Golden Syrian Hamster, Mouse, Pig, Primate, Rabbit, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.