Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF-1 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $452.71
Specifications
Antigen | IRF-1 |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABF188-25UG
|
MilliporeSigma
ABF18825UG |
25 μg |
Each for $192.35
|
|
|||||
ABF188100UG
|
MilliporeSigma
ABF188100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-IRF-1 is a rabbit polyclonal lantibody that detects Interferon regulatory factor 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.Specifications
IRF-1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Rat | |
IRF1 | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Rabbit | |
Inflammation and Immunology | |
NP_002189.1. | |
KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal half of human Interferon Regulatory Factor 1. | |
Affinity Purified | |
This rabbit polyclonal antibody detects Interferon Regulaotry Factor 1 in human cells. It targets an epitope within 14 amino acids from the N-terminal half. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title