Learn More
Description
Specifications
Specifications
| Antigen | IRF1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | interferon regulatory factor 1, MARIRF-1 |
| Gene Symbols | IRF1 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 200 and 250 of mouse Interferon Regulatory Factor 1 using the numbering given in entry NP_032416.1 (GeneID 16362). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
