Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF2 Antibody (JJ088-0), Novus Biologicals™
SDP

Catalog No. NBP267870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267870 Supplier Novus Biologicals Supplier No. NBP267870
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IRF2 Monoclonal specifically detects IRF2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IRF2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone JJ088-0
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias DKFZp686F0244, interferon regulatory factor 2, IRF-2
Gene Symbols IRF2
Host Species Rabbit
Immunogen Synthetic peptide within C-terminal human IRF2. (SwissProt: P14316 Human; SwissProt: P23906 Mouse)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3660
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.