Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF2BP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155309 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155309 Supplier Novus Biologicals Supplier No. NBP155309
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRF2BP1 Polyclonal specifically detects IRF2BP1 in Human samples. It is validated for Western Blot.

Specifications

Antigen IRF2BP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8IU81
Gene Alias DKFZP434M154, interferon regulatory factor 2 binding protein 1, interferon regulatory factor 2-binding protein 1, IRF-2-binding protein 1, IRF-2BP1
Gene Symbols IRF2BP1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to IRF2BP1(interferon regulatory factor 2 binding protein 1) The peptide sequence was selected from the middle region of IRF2BP1. Peptide sequence LVARNGEAEVSPTAGAEAVSGGGSGTGATPGAPLCCTLCRERLEDTHFVQ.
Molecular Weight of Antigen 62 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26145
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.