Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF2BP2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB341568 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341568 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341568 Supplier Novus Biologicals Supplier No. NBP281995PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRF2BP2 Polyclonal antibody specifically detects IRF2BP2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen IRF2BP2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias interferon regulatory factor 2 binding protein 2, IRF-2-binding protein 2
Host Species Rabbit
Immunogen IRF2BP2 antibody was raised against an 17 amino acid synthetic peptide near the amino terminus of human IRF2BP2. The immunogen is located within amino acids 150 - 200 of IRF2BP2. Amino Acid Squence: KLEEPPELNRQSPNPRR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359948
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.