Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF3 Antibody (3F10), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP104308Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104308Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104308Z Supplier Novus Biologicals Supplier No. NBP104308AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IRF3 Monoclonal specifically detects IRF3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Knockout Validated.

Specifications

Antigen IRF3
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Clone 3F10
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Knockout Validated
Gene Accession No. NP_001562
Gene Alias interferon regulatory factor 3, IRF-3
Gene Symbols IRF3
Host Species Mouse
Immunogen Recombinant human IRF-3(108-166aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3661
Test Specificity Anti-human IRF-3 mAb, clone 3F10, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human IRF3 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.