Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB195234 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB195234 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB195234 Supplier Novus Biologicals Supplier No. NBP176597F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRF3 Polyclonal antibody specifically detects IRF3 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen IRF3
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias interferon regulatory factor 3, IRF-3
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human IRF3. The immunogen is located within amino acids 330 - 380 of IRF3. Amino Acid Squence: GESWPQDQPWTKRL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3661
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.