Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF3 Antibody (PCRP-IRF3-3B2), FITC, Novus Biologicals™
SDP

Catalog No. NB154910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154910 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB154910 Supplier Novus Biologicals Supplier No. NBP314106F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Immunofluorescence

Specifications

Antigen IRF3
Applications Immunofluorescence
Classification Monoclonal
Clone PCRP-IRF3-3B2
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias interferon regulatory factor 3, IRF-3
Host Species Mouse
Immunogen Recombinant full-length human IRF3 protein (Uniprot: Q14653)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3661
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.