Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF4 Antibody (SR1924), Novus Biologicals™
SDP

Catalog No. NB260184 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260184 50 μg
NB259884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260184 Supplier Novus Biologicals Supplier No. NBP32182050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IRF4 Monoclonal antibody specifically detects IRF4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation

Specifications

Antigen IRF4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SR1924
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Flow Cytometry 1:120, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:100
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5
Host Species Rabbit
Immunogen A synthesized peptide derived from human IRF4 (Uniprot #: Q15306)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3662
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.