Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF5 Antibody (903430), PE, Novus Biologicals™
SDP

Catalog No. NB302567 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302567 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302567 Supplier Novus Biologicals Supplier No. FAB8447P

Rat Monoclonal Antibody

IRF5 Monoclonal antibody specifically detects IRF5 in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen IRF5
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 903430
Conjugate PE
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Host Species Rat
Immunogen E. coli-derived recombinant mouse IRF5, Ala208-Phe288, Accession # P56477
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3663
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.