Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF5 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001092 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001092 Supplier Novus Biologicals Supplier No. NB1001092
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 9 publications

IRF5 Polyclonal specifically detects IRF5 in Human, Mouse samples. It is validated for Chemotaxis, ChIP Assay, Flow Cytometry, KnockDown, Peptide ELISA, Western Blot.

Specifications

Antigen IRF5
Applications Chemotaxis, ChIP Assay, Flow Cytometry, KnockDown, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/mL, Flow Cytometry, Peptide ELISA Detection limit 1:128000, Chromatin Immunoprecipitation (ChIP)
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q13568
Gene Alias IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Gene Symbols IRF5
Host Species Goat
Immunogen Peptide with sequence C-QGPWPMHPAGMQ corresponding to C-Terminus according to NP_002191.1, NP_116032.1, NP_001092097.1, NP_001092099.1.
Molecular Weight of Antigen 56 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3663
Test Specificity This antibody is expected to recognize all four reported isoforms (NP_002191.1; NP_116032.1; NP_001092097.1; NP_001092099.1). Reported variants NP_001092100.1, NP_001092101.1, NP_001092098.1, NP_001092097.1 represent identical protein: Reported variants NP_116032.1, NP_001092100.1 represent identical protein
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.