Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF7 Antibody (3D9), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP104309G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104309G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104309G Supplier Novus Biologicals Supplier No. NBP104309G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IRF7 Monoclonal specifically detects IRF7 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IRF7
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3D9
Conjugate DyLight 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias interferon regulatory factor 7, interferon regulatory factor-7H, IRF-7, IRF7A, IRF-7H
Gene Symbols IRF7
Host Species Mouse
Immunogen Recombinant human IRF7 (1-150aa) purified from E. coli (NP_001563).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3665
Test Specificity Anti-human IRF7 mAb, clone 3D9, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human IRF7 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.