Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF7 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB180312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180312 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180312 Supplier Novus Biologicals Supplier No. NBP306987H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IRF7 Polyclonal antibody specifically detects IRF7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen IRF7
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias interferon regulatory factor 7, interferon regulatory factor-7H, IRF-7, IRF7A, IRF-7H
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the center of human IRF7. The immunogen is located within amino acids 200-230 of IRF7.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3665
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.