Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF9 Mouse anti-Human, Clone: PCRP-IRF9-2F8, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP31383520UG
This item is not returnable.
View return policy
Description
IRF9 Monoclonal antibody specifically detects IRF9 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein ArraySpecifications
IRF9 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array | |
Q00978 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Peptide Array | |
PCRP-IRF9-2F8 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma | |
Recombinant full-length human IRF9 protein (Uniprot: Q00978) | |
20 μg | |
Signal Transduction | |
10379 | |
Store at 4°C. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction