Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF9 Mouse anti-Human, Clone: PCRP-IRF9-2F8, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | IRF9 |
---|---|
Clone | PCRP-IRF9-2F8 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array |
Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
IRF9 Monoclonal antibody specifically detects IRF9 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein ArraySpecifications
IRF9 | |
0.2 mg/ml | |
Flow Cytometry, Immunofluorescence, Peptide Array | |
Unconjugated | |
Mouse | |
Signal Transduction | |
10 mM PBS with 0.05% BSA | |
IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma | |
Recombinant full-length human IRF9 protein (Uniprot: Q00978) | |
Primary | |
Store at 4°C. |
PCRP-IRF9-2F8 | |
Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q00978 | |
10379 | |
IgG2a | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title