Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium(IV) oxide powder, 99%

Catalog No. AAA1784903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1784903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1784903 Supplier Thermo Scientific Chemicals Supplier No. A1784903
Only null left
Add to Cart

CAS: 12030-49-8 | CH4 | 16.04 g/mol

Iridium(IV) oxide is used in the coating of anode-electrodes for industrial electrolysis in association with other rare oxides. It is also used in microelectrodes for electrophysiology research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iridium(IV) oxide is used in the coating of anode-electrodes for industrial electrolysis in association with other rare oxides. It is also used in microelectrodes for electrophysiology research.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-49-8
Molecular Formula IrO2
Molecular Weight (g/mol) 224.22
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD31654066 MFCD06411993 MFCD07370731
InChI Key HTXDPTMKBJXEOW-UHFFFAOYSA-N
IUPAC Name dioxoiridium
SMILES O=[Ir]=O

Specifications

Quantity 1 g
UN Number UN1479
Solubility Information Insoluble in water.
Formula Weight 224.2
Percent Purity 99%
Odor Odorless
Density 11.7
Chemical Name or Material Iridium(IV) oxide powder
Hazard Category H272-H315-H319
Hazard Statement GHS H Statement
H272
May intensify fire; oxidizer.
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 234-743-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.