Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium sponge, -100 mesh, 99.95% (metals basis)

Catalog No. aa1206904
Click to view available options
Quantity:
2 g

CAS: 7439-88-5 | Ir | 192.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Quantity 2 g
Mesh Size −100 Mesh
Form Sponge
UN Number UN3089
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Iridium sponge
Hazard Category H228-H319-H500
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 231-095-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.