Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Irisin/FNDC5 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB336906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336906 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336906 Supplier Novus Biologicals Supplier No. NBP259680MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Irisin/FNDC5 Polyclonal antibody specifically detects Irisin/FNDC5 in Human, Mouse, Camel samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Irisin/FNDC5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FRCP2Fibronectin type III repeat-containing protein 2
Host Species Rabbit
Immunogen Partial recombinant protein made to an internal region of the human Irisin/FNDC5 protein (amino acids 32-143) [UniProt Q8NAU1]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 252995
Target Species Human, Mouse, Camelid
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.