Iron granules, 99.98% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 3970830

Quantity 250g

 View more versions of this product

Catalog No. AA3970830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-89-6
    Physical Form Granules, 1-2mm (0.04-0.08 in.)
    Assay Percent Range 99.98% (Metals basis)
    Molecular Formula Fe
    MDL Number MFCD00010999
    Chemical Name or Material Iron

    Specifications

    Quantity 250g
    Color Gray
    Density 7.87g/cm3 at 20°C
    EINECS Number 231-096-4
    Physical Form Granules, 1-2mm (0.04-0.08 in.)
    Assay Percent Range 99.98% (Metals basis)
    Molecular Formula Fe
    CAS 7439-89-6
    Appearance Gray color granules
    MDL Number MFCD00010999
    Health Hazard 1 H319-H335: Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Iron

    H319-H335: Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit