Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) bromide, anhydrous, 98% min

Catalog No. AA3115406
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g

CAS: 7789-46-0 | Br2Fe | 215.65 g/mol

Iron(II) bromide is used as an intermediate in the synthesis porphryins. It is also used as weak reducing agents. It is used as precursor for the preparation of [(C2H5)4N]2FeBr4 by reacting with tetraethylammonium bromide. It is also involved in the synthesis of iron pyrite nanocrystals and nanocrystalline thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-46-0
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
IUPAC Name λ²-iron(2+) dibromide
SMILES [Fe++].[Br-].[Br-]

Specifications

Melting Point 684°C
Color Yellow
Physical Form Granules
Quantity 5 g
Assay Percent Range 98% min.
Solubility Information Soluble in water,methanol,ethanol and tetrahydrofuran.
Formula Weight 215.67
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Yellow granules
Density 4.636 g/cm3
Chemical Name or Material Iron(II) bromide, anhydrous
Show More Show Less
missing translation for 'healthHazard1' H314-H318
missing translation for 'healthHazard3' P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packing Group: III
EINECSNumber 232-168-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.