Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) oxalate hydrate, 96%

Catalog No. p-7050673
Click to view available options
Quantity:
500 g
2 kg

CAS: 516-03-0 | C2FeO4 | 143.86 g/mol

Used as a photographic developer, glass tint, decorative glass colorant, and pigment in plastics, paints, and lacquers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-03-0
Molecular Formula C2FeO4
Molecular Weight (g/mol) 143.86
MDL Number MFCD00012475
InChI Key OWZIYWAUNZMLRT-UHFFFAOYSA-L
Synonym ferrous oxalate, iron ii oxalate, unii-dzp4yv3icv, dzp4yv3icv, iron protoxalate, iron, ethanedioato 2--.kappa.o1,.kappa.o2, iron 2+ oxalate, iron oxalate fec2o4, hsdb 463, iron 2+ ; oxalate
PubChem CID 10589
IUPAC Name iron(2+);oxalate
SMILES [Fe++].[O-]C(=O)C([O-])=O

Specifications

Melting Point 190°C (decomposition)
Density 2.28 g/mL
Quantity 500 g
UN Number UN3288
Sensitivity Hygroscopic
Solubility Information Soluble in acids
Formula Weight 143.87 (Anhydrous)
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Iron(II) oxalate hydrate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 208-217-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.