Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) bromide, 90+%

Catalog No. AC384471000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC384471000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC384471000 Supplier Thermo Scientific Chemicals Supplier No. 384471000

CAS: 10031-26-2 | Br3Fe | 295.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-26-2
Molecular Formula Br3Fe
Molecular Weight (g/mol) 295.56
MDL Number MFCD00016082

Specifications

Melting Point 200°C
Color Beige-Yellow to White
Physical Form Powder
Quantity 100 g
Assay Percent Range 90% min. (Iodometry)
Linear Formula FeBr3
Merck Index 15, 4045
Solubility Information Solubility in water: soluble
Formula Weight 295.56
Percent Purity ≥90%
Packaging Glass bottle
Chemical Name or Material Iron(III) bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
EINECSNumber 233-089-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.