Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) bromide, anhydrous, 98+%

Catalog No. p-7049913
Click to view available options
Quantity:
5 g
100 g

CAS: 10031-26-2

Iron(III) bromide is used as a Lewis acid catalyst in the halogenation of aromatic compounds. It is also used in the preparation of 2,5-dibromo-4-aryl-1,3-pentadiene by reacting with homoallenic bromohydrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-26-2
MDL Number MFCD00016082

Specifications

Melting Point (decomposition)
Color Red
Physical Form Crystalline
Quantity 5 g
Merck Index 14,4018
Solubility Information Soluble in water,ether and alcohol.
Formula Weight 295.57
Percent Purity ≥98%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Iron(III) bromide, Anhydrous
Show More Show Less
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H314-H302-H335-H319
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-089-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.