Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron powder, -200 mesh, 99+% (metals basis), Thermo Scientific Chemicals

Catalog No. AA00737A1
Click to view available options
Quantity:
250 g
1 kg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Density 7.87
Quantity 1 kg
Mesh Size -200 Mesh
Form Powder
Assay Percent Range 99+% (metals basis)
UN Number UN3089
Formula Weight 55.847
Percent Purity ≥99%
Assay (metals basis)
Chemical Name or Material Iron
Hazard Category H500-X
Hazard Statement GHS H Statement
H228-H319-H335
Precautionary Statement P235+P410-P280
DOTInformation Hazard Class: 4.2; Packaging Group: II
EINECSNumber 231-096-4
RTECSNumber NO4565500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.