Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iroquois-class homeodomain protein IRX-5 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB350292 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350292 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350292 Supplier Novus Biologicals Supplier No. NBP321441APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Iroquois-class homeodomain protein IRX-5 Polyclonal antibody specifically detects Iroquois-class homeodomain protein IRX-5 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Iroquois-class homeodomain protein IRX-5
Applications Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias Homeodomain protein IRX-2A, Homeodomain protein IRXB2, iroquois homeobox 5, Iroquois homeobox protein 5, iroquois-class homeodomain protein IRX-5, IRX-2a, IRX2A, IRXB2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Iroquois-class homeodomain protein IRX-5 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10265
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.