Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRP2 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB1001798C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001798C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001798C Supplier Novus Biologicals Supplier No. NB1001798C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRP2 Polyclonal specifically detects IRP2 in Human, Mouse samples. It is validated for Gel Super Shift Assays, Gel Supershift, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IRP2
Applications Gel Supershift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ACO3, EC 4.2.1.3, FLJ23381, IRE-BP 2, Iron regulatory protein 2, iron-responsive element binding protein 2, iron-responsive element-binding protein 2, IRP2IRP2AD
Gene Symbols IREB2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the murine IRP2 protein sequence (between residues 100-200). [UniProt Q811J3]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline HIF Target Genes, Hypoxia, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3658
Test Specificity Mouse. Preliminary results have shown poor reactivity against human protein. Immunogen has 94% homology with human, primate and rat proteins.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.