Learn More
Thermco Products Irreversible Freeze Indicator (0°C/32°F)
Catalog No. 04500361
Change view
Click to view available options
Model:
Indicator A
Indicator B
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
04-500-361 Indicator A
04-500-362 Indicator B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-500-361 Supplier Thermco Products Supplier No. F1032G2
Only null left
Add to Cart
Add to Cart
Self-adhesive labels / Irreversible color change - Green to Red / Self-activating / Storage: Room Temperature
  • The Freeze Check Indicator is a self-activating temperature indicator that displays an irreversible chemical color change if the temperature drops below 0°C/32°F.
  • Durable, low-profile design adheres to various surfaces, making it ideal for pharmaceuticals, food, and industrial goods.
  • No pre-activation is needed, simply apply the self-adhesive label.
Specifications
Material Paper
Model Indicator A
Product Type Chemical Temperature Indicator