Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS1 Antibody (OTI3G10), HRP, Novus Biologicals™
SDP

Catalog No. NBP271049H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271049H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271049H Supplier Novus Biologicals Supplier No. NBP271049H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IRS1 Monoclonal specifically detects IRS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen IRS1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3G10
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias HIRS-1, insulin receptor substrate 1, IRS-1
Gene Symbols IRS1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-280 of human IRS1 (NP_005535) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Nutrient Sensing in the Brain, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3667.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.