Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS1, p Ser636 Antibody, Novus Biologicals™
SDP

Catalog No. NB017855 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017855 0.025 mL
NB10082003 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017855 Supplier Novus Biologicals Supplier No. NB100820030.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRS1 (p Ser636) Polyclonal specifically detects IRS1 (p Ser636) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IRS1 (p Ser636)
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:100
Formulation phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Gene Accession No. P35568
Gene Alias HIRS-1, insulin receptor substrate 1, IRS-1
Gene Symbols IRS1
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human IRS-1 around the phosphorylation site of serine 636 (P-M-SP-P-K).
Molecular Weight of Antigen 180 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Diabetes Research, mTOR Pathway, Nutrient Sensing in the Brain, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3667
Test Specificity IRS-1 (specific to Phospho-Ser636) detects endogenous levels of IRS-1 only when phosphorylated at serine 636.
Target Species Human, Rat, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.