Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS1, p Tyr1179 Antibody, Novus Biologicals™
SDP

Catalog No. NB10074450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074450 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074450 Supplier Novus Biologicals Supplier No. NB10074450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRS1 (p Tyr1179) Polyclonal specifically detects IRS1 (p Tyr1179) in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IRS1 (p Tyr1179)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000
Formulation PBS with 1mg/ml BSA with 0.05% Sodium Azide
Gene Alias HIRS-1, insulin receptor substrate 1, IRS-1
Gene Symbols IRS1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues L(1173) E N G L N (pY) I D L D L V K(1186) of human IRS-1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3667
Test Specificity Detects phosphorylated IRS-1 (Tyr1173) in mouse cells.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.