Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Ischemin sodium salt
Click to view available options
Quantity:
10 mg
50 mg
Description
Ischemin sodium salt is a CBP bromodomain inhibitor. Inhibits p53 interaction with CBP and decreases p53 transcriptional activity. Protects against Doxorubicin -induced apoptosis in cardiomyocytes in vitro. Cell permeable.
Specifications
Specifications
| Purity | >99% |
| Quantity | 10 mg |
| Product Type | Ischemin sodium salt |
| Molecular Weight (g/mol) | 357.36 |
| Inhibitors | Ischemin sodium salt |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction