Learn More
World Precision Instrument ISDB Blank Panels
SDP
Catalog No. 502521231
Encompass
The ISDB chassis (74030) holds up to eight modules and includes a power supply. You may include any combination of BRIDGE8 Transducer Amplifier modules or ISO-DAM8A single channel modules (74020). If you choose fewer than eight modules, you need to order the ISDB blank panels (74050) to fill the empty slots in the chassis. Please specify line voltage when ordering.
Catalog No. 50-252-1231 Supplier World Precision Instrument Supplier No. 74050
The ISDB chassis (74030) holds up to eight modules and includes a power supply. You may include any combination of BRIDGE8 Transducer Amplifier modules or ISO-DAM8A single channel modules (74020). If you choose fewer than eight modules, you need to order the ISDB blank panels (74050) to fill the empty slots in the chassis. Please specify line voltage when ordering.