Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISG15/UCRP Rabbit anti-Human, Clone: S08-1H5, Novus Biologicals™
SDP

Catalog No. NB188250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188250 100 μg
NB188251 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188250 Supplier Novus Biologicals Supplier No. NBP314947100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ISG15/UCRP Monoclonal antibody specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ISG15/UCRP
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S08-1H5
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias G1P2interferon-induced 17-kDa/15-kDa protein, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRPIP17
Host Species Rabbit
Immunogen A synthetic peptide of human ISG15/UCRP (Uniprot#: P05161)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 9636
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.