Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Islet-1 Antibody (OTI2D8), PerCP, Novus Biologicals™
SDP

Catalog No. NB289137 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289137 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289137 Supplier Novus Biologicals Supplier No. NBP271050PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Islet-1 Monoclonal antibody specifically detects Islet-1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Islet-1
Applications Western Blot
Classification Monoclonal
Clone OTI2D8
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias insulin gene enhancer protein ISL-1, ISL LIM homeobox 1, Isl-1, ISL1 transcription factor, LIM/homeodomain, ISL1 transcription factor, LIM/homeodomain, (islet-1), Islet-1, ISLET1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 134-349 of human ISL1(NP_002193) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3670
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.