Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ISLR Monoclonal Antibody (C3), Invitrogen™
Mouse Monoclonal Antibody
$243.00 - $522.50
Specifications
Antigen | ISLR |
---|---|
Clone | C3 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543685
|
Invitrogen™
MA543685 |
20 μL |
Each for $243.00
|
|
|||||
PIMA543684
|
Invitrogen™
MA543684 |
100 μL |
Each for $522.50
|
|
|||||
Description
ISLR is a protein coding gene.Specifications
ISLR | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
HsT17563; immunoglobulin superfamily containing leucine rich repeat; immunoglobulin superfamily containing leucine-rich repeat protein; ISLR; Meflin; mesenchymal stromal cell- and fibroblast-expressing Linx paralogue; UNQ189/PRO215 | |
ISLR | |
IgG1 κ | |
Protein A/G | |
Antibody |
C3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O14498 | |
3671 | |
Cys19-Phe428 (Accession O14498), with N-terminal His Tag. | |
Primary | |
-20°C | |
ISLR |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title