Learn More
Isobutanol, 99%
Description
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for the liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful in the production of renewable chemicals and fuels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for the liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful in the production of renewable chemicals and fuels.
Solubility
Miscible with alcohol and diethyl ether. Partially miscible with water.
Notes
Avoid all sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
| pH | 7 |
| Melting Point | -108°C |
| Density | 0.802 |
| Boiling Point | 107°C to 108°C |
| Quantity | 2500 mL |
| Flash Point | 28°C (82°F) |
| Odor | Sweet |
| Refractive Index | 1.396 |
| Linear Formula | (CH3)2CHCH2OH |
| UN Number | UN1212 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H318-H335-H336 |
| Hazard Statement | GHS H Statement H318-H226-H335-H336-H315 Causes serious eye damage. Flammable liquid and vapour. May cause respiratory irritation. May cause drowsiness or dizziness. Causes skin irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTANOL |
| EINECSNumber | 201-148-0 |
| RTECSNumber | NP9625000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.