Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyrophenone, 97%

Catalog No. p-7027684
Click to view available options
Quantity:
25 g
100 g

CAS: 611-70-1 | C10H12O | 148.21 g/mol

Isobutyrophenone is used as an internal standard in the determination of hydroxyzine hydrochloride and benzyl alcohol in injection solutions. It is also used in the preparation of alpha-hydroxyisobutyrophenone. Isobutyrophenone is employed as photosensitizer intermediate. It can react with bromoacetic acid ethyl ester to get 3-hydroxy-4-methyl-3-phenyl-valeric acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyrophenone is used as an internal standard in the determination of hydroxyzine hydrochloride and benzyl alcohol in injection solutions. It is also used in the preparation of alpha-hydroxyisobutyrophenone. Isobutyrophenone is employed as photosensitizer intermediate. It can react with bromoacetic acid ethyl ester to get 3-hydroxy-4-methyl-3-phenyl-valeric acid ethyl ester.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-70-1
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00008917
InChI Key BSMGLVDZZMBWQB-UHFFFAOYSA-N
Synonym isobutyrophenone, isopropyl phenyl ketone, 2-methylpropiophenone, 1-propanone, 2-methyl-1-phenyl, phenyl isopropyl ketone, isopropyl phenyl keton, alpha-methylpropiophenone, unii-8l53972ntz, .alpha.-methylpropiophenone, 2-methyl-1-phenyl-1-propanone
PubChem CID 69144
IUPAC Name 2-methyl-1-phenylpropan-1-one
SMILES CC(C)C(=O)C1=CC=CC=C1

Specifications

Melting Point 1°C
Density 0.988
Boiling Point 217°C
Flash Point 84°C (184°F)
Refractive Index 1.517
Quantity 25 g
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 148.21
Percent Purity 97%
Chemical Name or Material Isobutyrophenone
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-275-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.