Learn More
Invitrogen™ ISOC1 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Specifications
Specifications
| Antigen | ISOC1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ISOC1 |
| Gene Accession No. | Q96CN7 |
| Gene Alias | 2610034N03Rik; CGI-111; Down-regulated in nephrectomized rat kidney #1; down-regulated in nephrectomized rat kidney protein 1; DR-NR#1; DR-NR1; isoc1; isochorismatase domain containing 1; isochorismatase domain-containing protein 1; isochorismatase domain-containing protein 1; LOW QUALITY PROTEIN: isochorismatase domain-containing protein 1; RGD1307632; zgc:153699 |
| Gene Symbols | ISOC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.