Learn More
Description
Specifications
Specifications
| Antigen | Isocitrate Dehydrogenase 1/IDH1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown |
| Classification | Polyclonal |
| Concentration | 1.0mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:10000, Immunohistochemistry 10 - 1:500, Immunoprecipitation 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Knockout Validated, Knockdown Validated |
| Formulation | PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 |
| Gene Alias | Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD |
| Gene Symbols | IDH1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
